Assay Design



Gene Transcripts

Gene Symbol
Tceal5
Entrez Gene ID
331532
Gene Name
transcription elongation factor A (SII)-like 5
Gene Aliases
6430401A05Rik
Chromosome Location
Chr.X: 136200948 - 136203851 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177919.2 NP_808587.1 1 - 2 86 79
GenBank mRNA AK032140.1 - 1 - 2 88 79
BC100415.1 - 1 - 2 111 79

View Full Product Details