Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007465.3 | NP_001007466.1 | 1 - 2 | 200 | 94 |
NM_001164571.1 | NP_001158043.1 | 1 - 2 | 200 | 94 | |
GenBank mRNA | AK150213.1 | - | 1 - 2 | 133 | 94 |
AK159442.1 | - | 1 - 2 | 159 | 94 | |
AK169393.1 | - | 1 - 2 | 141 | 94 | |
AK169988.1 | - | 1 - 2 | 201 | 94 |