Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021556.3 | NP_067531.1 | 2 - 3 | 753 | 82 |
GenBank mRNA | AK011476.1 | - | 2 - 3 | 755 | 82 |
AK013360.1 | - | 2 - 3 | 589 | 82 | |
AK151642.1 | - | 2 - 3 | 754 | 82 | |
BC029784.1 | - | 2 - 3 | 751 | 82 |