Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172149.5 | NP_742161.4 | 3 - 4 | 285 | 103 |
GenBank mRNA | AY547731.1 | - | 3 - 4 | 271 | 103 |
BC086888.1 | - | 3 - 4 | 274 | 103 | |
CV560804.1 | - | 3 - 4 | 286 | 103 |