Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171034.1 | NP_001164505.1 | 3 - 4 | 394 | 106 |
NM_001171035.1 | NP_001164506.1 | 2 - 3 | 250 | 106 | |
NM_001171036.1 | NP_001164507.1 | 3 - 4 | 215 | 106 | |
NM_026669.4 | NP_080945.1 | 2 - 3 | 179 | 106 | |
GenBank mRNA | AK019865.2 | - | 2 - 3 | 141 | 106 |
AK036782.1 | - | 2 - 3 | 141 | 106 | |
AK036994.1 | - | 2 - 3 | 141 | 106 | |
AK049604.1 | - | 2 - 3 | 130 | 106 | |
AK050299.1 | - | 2 - 3 | 142 | 106 | |
AK050323.1 | - | 2 - 3 | 130 | 106 | |
AK088030.1 | - | 2 - 3 | 141 | 106 | |
AK088683.1 | - | 2 - 3 | 142 | 106 | |
AK139901.1 | - | 2 - 3 | 142 | 106 | |
AK159380.1 | - | 2 - 3 | 141 | 106 | |
AK166935.1 | - | 2 - 3 | 141 | 106 | |
AK187051.1 | - | 1 - 2 | 86 | 106 | |
AK208219.1 | - | 1 - 2 | 85 | 106 | |
AK217190.1 | - | 2 - 3 | 105 | 106 | |
BC005588.1 | - | 2 - 3 | 124 | 106 | |
BY272301.1 | - | 2 - 3 | 180 | 106 | |
BY753376.1 | - | 2 - 3 | 233 | 106 | |
CD554385.1 | - | 3 - 4 | 334 | 106 |