Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009029.2 | NP_033055.2 | 3 - 4 | 514 | 119 |
GenBank mRNA | AK011246.1 | - | 3 - 4 | 514 | 119 |
BC096525.1 | - | 3 - 4 | 514 | 119 | |
DQ400415.1 | - | 3 - 4 | 364 | 119 | |
M26391.1 | - | 3 - 4 | 462 | 119 |