Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170694.1 | NP_001164165.1 | 11 - 12 | 1398 | 70 |
NM_134083.4 | NP_598844.3 | 11 - 12 | 1393 | 70 | |
NR_033185.1 | 12 - 13 | 1778 | 70 | ||
GenBank mRNA | AK013540.1 | - | - | 1503 | 70 |
AK031242.1 | - | 11 - 12 | 1359 | 70 | |
AK031248.1 | - | 11 - 12 | 1364 | 70 | |
AK147784.1 | - | 12 - 13 | 1399 | 70 | |
AK160881.1 | - | 12 - 13 | 1585 | 70 | |
BC003224.1 | - | 9 - 10 | 1191 | 70 | |
JN086338.1 | - | 10 - 11 | 1314 | 70 | |
JN086339.1 | - | 11 - 12 | 1552 | 70 |