Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170694.1 | NP_001164165.1 | 12 - 13 | 1529 | 147 |
NM_134083.4 | NP_598844.3 | 12 - 13 | 1524 | 147 | |
NR_033185.1 | 13 - 14 | 1909 | 147 | ||
GenBank mRNA | AK013540.1 | - | - | 1634 | 147 |
AK031242.1 | - | 12 - 13 | 1490 | 147 | |
AK031248.1 | - | 12 - 13 | 1495 | 147 | |
AK147784.1 | - | 13 - 14 | 1530 | 147 | |
AK160881.1 | - | 13 - 14 | 1716 | 147 | |
AK191301.1 | - | 1 - 2 | 81 | 147 | |
AK204057.1 | - | 2 - 3 | 147 | ||
AK210358.1 | - | 2 - 3 | 159 | 147 | |
AK217036.1 | - | 2 - 3 | 159 | 147 | |
BC003224.1 | - | 10 - 11 | 1322 | 147 | |
JN086338.1 | - | 11 - 12 | 1445 | 147 | |
JN086339.1 | - | 12 - 13 | 1683 | 147 |