Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177381.3 | NP_796355.2 | 17 - 18 | 1985 | 100 |
GenBank mRNA | BC038030.1 | - | 18 - 19 | 1996 | 100 |
BC048005.1 | - | 13 - 14 | 1340 | 100 | |
CO432313.1 | - | 4 - 5 | 421 | 100 |