Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025959.3 | NP_080235.2 | 13 - 14 | 1076 | 71 |
GenBank mRNA | AK008996.1 | - | - | 820 | 71 |
AK012174.1 | - | 13 - 14 | 1077 | 71 | |
AK014354.1 | - | 13 - 14 | 1064 | 71 | |
AK144728.1 | - | - | 1073 | 71 | |
AK166961.1 | - | 13 - 14 | 1061 | 71 | |
BC025134.1 | - | 9 - 10 | 764 | 71 | |
BC037099.1 | - | 13 - 14 | 1074 | 71 | |
BC043044.1 | - | - | 1055 | 71 | |
BC057997.1 | - | - | 1032 | 71 | |
BC116752.1 | - | 13 - 14 | 1076 | 71 | |
BC119168.1 | - | 13 - 14 | 1076 | 71 |