Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009982.4 | NP_034112.3 | 2 - 3 | 435 | 87 |
GenBank mRNA | AK002454.1 | - | 2 - 3 | 394 | 87 |
AK049406.1 | - | 2 - 3 | 394 | 87 | |
AK146858.1 | - | 2 - 3 | 443 | 87 | |
AK150761.1 | - | 2 - 3 | 395 | 87 | |
AK151388.1 | - | 2 - 3 | 394 | 87 | |
AK151711.1 | - | 2 - 3 | 395 | 87 | |
AK151861.1 | - | 2 - 3 | 281 | 87 | |
AK153168.1 | - | 2 - 3 | 395 | 87 | |
AK167882.1 | - | 2 - 3 | 394 | 87 | |
BC067063.1 | - | 2 - 3 | 368 | 87 | |
U74683.3 | - | 2 - 3 | 392 | 87 | |
U89269.1 | - | 2 - 3 | 393 | 87 |