Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026033.2 | NP_080309.2 | 1 - 2 | 555 | 71 |
GenBank mRNA | AB047921.1 | - | 1 - 2 | 536 | 71 |
AK012921.1 | - | 1 - 2 | 556 | 71 | |
AK078416.1 | - | 1 - 2 | 500 | 71 | |
BC019449.1 | - | 1 - 2 | 425 | 71 |