Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007870.3 | NP_031896.3 | 4 - 5 | 646 | 79 |
GenBank mRNA | AF047355.1 | - | 4 - 5 | 621 | 79 |
AK156109.1 | - | - | 479 | 79 | |
AK171843.1 | - | 4 - 5 | 479 | 79 | |
AK172254.1 | - | 4 - 5 | 1039 | 79 | |
BC012671.1 | - | 4 - 5 | 599 | 79 | |
U76110.1 | - | 4 - 5 | 455 | 79 |