Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289568.1 | NP_001276497.1 | 16 - 17 | 2005 | 94 |
NM_001289571.1 | NP_001276500.1 | 14 - 15 | 1861 | 94 | |
NM_028349.3 | NP_082625.2 | 15 - 16 | 1922 | 94 | |
GenBank mRNA | AK145076.1 | - | 14 - 15 | 1810 | 94 |
AK162883.1 | - | 1 - 2 | 1283 | 94 | |
AK165224.1 | - | 16 - 17 | 1951 | 94 |