Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113478.1 | NP_001106950.1 | 4 - 5 | 511 | 93 |
NM_009146.3 | NP_033172.2 | 4 - 5 | 473 | 93 | |
GenBank mRNA | AK075573.1 | - | 4 - 5 | 465 | 93 |
AK144933.1 | - | 4 - 5 | 465 | 93 | |
AK162693.1 | - | 4 - 5 | 466 | 93 | |
AK167289.1 | - | 4 - 5 | 501 | 93 | |
AU067508.1 | - | 4 - 5 | 473 | 93 | |
BC027770.1 | - | 3 - 4 | 471 | 93 | |
D50464.1 | - | 4 - 5 | 461 | 93 |