Assay Design
Gene Transcripts
- Gene Symbol
- Otud7a
- Entrez Gene ID
-
170711
- Gene Name
- OTU domain containing 7A
- Gene Aliases
-
AJ430384, Otud7
- Chromosome Location
-
Chr.7: 63444745 - 63759905 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_130880.1 |
NP_570950.1 |
7 - 8 |
1045 |
67 |
GenBank mRNA |
AJ430384.1 |
- |
7 - 8 |
1045 |
67 |
View Full Product Details