Assay Design



Gene Transcripts

Gene Symbol
Unc5c
Entrez Gene ID
22253
Gene Name
unc-5 homolog C (C. elegans)
Gene Aliases
B130051O18Rik, Unc5h3, rcm
Chromosome Location
Chr.3: 141465564 - 141834924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293561.1 NP_001280490.1 16 - 17 2838 70
NM_009472.4 NP_033498.1 15 - 16 2781 70
GenBank mRNA AK031655.1 - 16 - 17 2805 70
AK138987.1 - 4 - 5 578 70
AK141181.1 - 16 - 17 2804 70
AK142411.1 - 16 - 17 2804 70
AK142974.1 - 16 - 17 2732 70
AK164239.1 - 15 - 16 2582 70
BC115772.1 - 16 - 17 2828 70
U72634.1 - 15 - 16 2781 70

View Full Product Details