Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178654.4 | NP_848769.2 | 17 - 18 | 2678 | 97 |
GenBank mRNA | AK049713.1 | - | 17 - 18 | 2665 | 97 |
AK054281.1 | - | 3 - 4 | 282 | 97 | |
AK083425.1 | - | 12 - 13 | 1476 | 97 | |
AK133298.1 | - | 17 - 18 | 2667 | 97 | |
AK171092.1 | - | 17 - 18 | 2609 | 97 | |
BC052073.1 | - | 18 - 19 | 2601 | 97 |