Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028228.3 | NP_082504.1 | 5 - 6 | 504 | 139 |
GenBank mRNA | AF321817.1 | - | 5 - 6 | 452 | 139 |
AF421879.1 | - | 5 - 6 | 387 | 139 | |
AJ344106.1 | - | 4 - 5 | 276 | 139 | |
AK012057.1 | - | 5 - 6 | 482 | 139 | |
AK075840.1 | - | 3 - 4 | 274 | 139 | |
BC065781.1 | - | 5 - 6 | 403 | 139 | |
BC115636.1 | - | 5 - 6 | 416 | 139 | |
BC115637.1 | - | 5 - 6 | 414 | 139 |