Assay Design



Gene Transcripts

Gene Symbol
Prss51
Entrez Gene ID
100504162
Gene Name
protease, serine 51
Gene Aliases
1700007N14Rik
Chromosome Location
Chr.14: 64085525 - 64097674 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK005740.1 - 1 - 2 168 88

View Full Product Details