Assay Design



Gene Transcripts

Gene Symbol
Prss51
Entrez Gene ID
100504162
Gene Name
protease, serine 51
Gene Aliases
1700007N14Rik
Chromosome Location
Chr.14: 64085525 - 64097674 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193631.1 NP_001180560.1 4 - 5 648 72
GenBank mRNA AK005740.1 - 4 - 5 653 72
BC048657.1 - 4 - 5 648 72

View Full Product Details