Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153137.4 | NP_694777.3 | 2 - 3 | 355 | 72 |
GenBank mRNA | AK031400.1 | - | 2 - 3 | 295 | 72 |
AK036608.1 | - | 2 - 3 | 124 | 72 | |
AK041994.1 | - | 2 - 3 | 356 | 72 | |
AK080080.1 | - | 2 - 3 | 325 | 72 | |
BC036561.1 | - | 2 - 3 | 163 | 72 |