Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016851.2 | NP_058547.2 | 6 - 7 | 1250 | 72 |
GenBank mRNA | AK143299.1 | - | 6 - 7 | 1201 | 72 |
AK143954.1 | - | 6 - 7 | 1235 | 72 | |
AK149487.1 | - | 6 - 7 | 1213 | 72 | |
BC008515.1 | - | 6 - 7 | 1200 | 72 | |
U73029.1 | - | 6 - 7 | 1200 | 72 |