Assay Design



Gene Transcripts

Gene Symbol
Fam71a
Entrez Gene ID
619288
Gene Name
family with sequence similarity 71, member A
Gene Aliases
4933417M04Rik, GARI-L4
Chromosome Location
Chr.1: 191162584 - 191164817 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109759.1 NP_001103229.1 1 - 1 2163 94
GenBank mRNA AK133412.1 - 1 - 1 2165 94

View Full Product Details