Assay Design



Gene Transcripts

Gene Symbol
Spta1
Entrez Gene ID
20739
Gene Name
spectrin alpha, erythrocytic 1
Gene Aliases
AF093576, AI451697, Spna-1, Spna1, ha, ihj, nmf4, sph
Chromosome Location
Chr.1: 174172776 - 174248449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011465.4 NP_035595.2 6 - 7 973 128
GenBank mRNA AF093576.1 - 6 - 7 881 128
AK142860.1 - 6 - 7 948 128
BC150747.1 - 6 - 7 838 128
U87455.1 - 6 - 7 953 128

View Full Product Details