Assay Design



Gene Transcripts

Gene Symbol
Spta1
Entrez Gene ID
20739
Gene Name
spectrin alpha, erythrocytic 1
Gene Aliases
AF093576, AI451697, Spna-1, Spna1, ha, ihj, nmf4, sph
Chromosome Location
Chr.1: 174172776 - 174248449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011465.4 NP_035595.2 7 - 8 82
GenBank mRNA AF093576.1 - 7 - 8 82
AK142860.1 - 7 - 8 82
BC150747.1 - 7 - 8 977 82
U87455.1 - 7 - 8 1092 82

View Full Product Details