Assay Design



Gene Transcripts

Gene Symbol
Ocrl
Entrez Gene ID
320634
Gene Name
oculocerebrorenal syndrome of Lowe
Gene Aliases
9530014D17Rik, BB143339, OCRL1
Chromosome Location
Chr.X: 47912456 - 47966428 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177215.3 NP_796189.2 20 - 21 2451 96
GenBank mRNA AK041963.1 - 20 - 21 2453 96
BC043709.1 - 7 - 8 833 96
BC054840.1 - 1 - 2 282 96
BC068146.1 - 20 - 21 2451 96

View Full Product Details