Assay Design



Gene Transcripts

Gene Symbol
Mettl3
Entrez Gene ID
56335
Gene Name
methyltransferase like 3
Gene Aliases
2310024F18Rik, M6A, Spo8
Chromosome Location
Chr.14: 52294841 - 52305124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019721.2 NP_062695.2 4 - 5 1040 90
GenBank mRNA AK009492.1 - 1 - 2 101 90
AK085189.1 - 4 - 5 1027 90
AK151616.1 - 4 - 5 1027 90
AK153113.1 - 4 - 5 1027 90
BC012526.1 - 4 - 5 1024 90

View Full Product Details