Assay Design



Gene Transcripts

Gene Symbol
Gipr
Entrez Gene ID
381853
Gene Name
gastric inhibitory polypeptide receptor
Gene Aliases
GIP-R, Gm1081, Gm160
Chromosome Location
Chr.7: 19156063 - 19170248 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080815.1 NP_001074284.1 12 - 13 1303 92
GenBank mRNA AK133386.1 - 6 - 7 1157 92
AK146552.1 - 10 - 11 1635 92

View Full Product Details