Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019637.3 | NP_062611.2 | 7 - 8 | 669 | 64 |
GenBank mRNA | AK002822.1 | - | - | 611 | 64 |
AK030122.1 | - | 7 - 8 | 669 | 64 | |
AK042741.1 | - | 8 - 9 | 724 | 64 | |
AK046970.1 | - | 7 - 8 | 646 | 64 | |
AK154136.1 | - | 7 - 8 | 764 | 64 | |
AK166043.1 | - | 7 - 8 | 652 | 64 | |
BC038608.1 | - | 7 - 8 | 619 | 64 | |
BC096674.1 | - | 6 - 7 | 579 | 64 | |
U34973.1 | - | 7 - 8 | 637 | 64 |