Assay Design



Gene Transcripts

Gene Symbol
Gtpbp10
Entrez Gene ID
207704
Gene Name
GTP-binding protein 10 (putative)
Gene Aliases
4930545J22Rik, BC034507
Chromosome Location
Chr.5: 5537450 - 5559774 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153116.1 NP_694756.1 1 - 2 70 68
GenBank mRNA AK139595.1 - 1 - 2 84 68
AK160252.1 - 1 - 2 108 68
BC034507.1 - 1 - 2 96 68

View Full Product Details