Assay Design



Gene Transcripts

Gene Symbol
Krit1
Entrez Gene ID
79264
Gene Name
KRIT1, ankyrin repeat containing
Gene Aliases
2010007K12Rik, A630036P20Rik, AA432855, AI450393, AI643869, BB155247, BB235701, Ccm1
Chromosome Location
Chr.5: 3803165 - 3844515 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030675.3 NP_109600.2 1 - 2 320 96
GenBank mRNA AF310134.1 - 1 - 2 296 96
AK144667.1 - 1 - 2 302 96
BB619038.1 - 1 - 2 302 96
BB855141.1 - 1 - 2 320 96
BC016087.1 - 1 - 2 270 96
BC141042.1 - 1 - 2 164 96

View Full Product Details