Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170552.1 | NP_001164023.1 | 2 - 3 | 473 | 128 |
NM_030675.3 | NP_109600.2 | 2 - 3 | 427 | 128 | |
NR_033173.1 | 2 - 3 | 526 | 128 | ||
GenBank mRNA | AF306509.1 | - | 2 - 3 | 140 | 128 |
AF310134.1 | - | 2 - 3 | 403 | 128 | |
AK039264.1 | - | - | 448 | 128 | |
AK085107.1 | - | 2 - 3 | 455 | 128 | |
AK144667.1 | - | 2 - 3 | 409 | 128 | |
AY328895.1 | - | 1 - 2 | 109 | 128 | |
AY464945.1 | - | 1 - 2 | 109 | 128 | |
BB619038.1 | - | 2 - 3 | 409 | 128 | |
BC016087.1 | - | 2 - 3 | 377 | 128 | |
BC044820.1 | - | 1 - 2 | 168 | 128 | |
BC054819.1 | - | 2 - 3 | 518 | 128 | |
BC085252.1 | - | 2 - 3 | 233 | 128 | |
BC141042.1 | - | 2 - 3 | 271 | 128 |