Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170552.1 | NP_001164023.1 | 7 - 8 | 1211 | 76 |
NM_030675.3 | NP_109600.2 | 7 - 8 | 1165 | 76 | |
NR_033173.1 | 7 - 8 | 1264 | 76 | ||
GenBank mRNA | AF310134.1 | - | 7 - 8 | 1141 | 76 |
AK085107.1 | - | 7 - 8 | 1193 | 76 | |
AK144667.1 | - | 7 - 8 | 1147 | 76 | |
AY328895.1 | - | 6 - 7 | 847 | 76 | |
AY464945.1 | - | 6 - 7 | 847 | 76 | |
BC016087.1 | - | 7 - 8 | 1115 | 76 | |
BC044820.1 | - | 6 - 7 | 906 | 76 | |
BC054819.1 | - | 7 - 8 | 1256 | 76 | |
BC085252.1 | - | 7 - 8 | 971 | 76 | |
BC141042.1 | - | 7 - 8 | 1009 | 76 |