Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019550.2 | NP_062423.1 | 2 - 3 | 218 | 64 |
GenBank mRNA | AF095718.2 | - | 2 - 3 | 88 | 64 |
AK015056.1 | - | 2 - 3 | 138 | 64 | |
AK041655.1 | - | 2 - 3 | 117 | 64 | |
AK050624.1 | - | 2 - 3 | 99 | 64 | |
AK050650.1 | - | 2 - 3 | 99 | 64 | |
AK135738.1 | - | 2 - 3 | 219 | 64 | |
AK137348.1 | - | 2 - 3 | 97 | 64 | |
AK207246.1 | - | 2 - 3 | 49 | 64 | |
BC010255.1 | - | 2 - 3 | 104 | 64 |