Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017381.2 | NP_059077.1 | 8 - 9 | 798 | 99 |
GenBank mRNA | AF133818.1 | - | 8 - 9 | 98 | |
AK146275.1 | - | 8 - 9 | 796 | 99 | |
AK146854.1 | - | 8 - 9 | 790 | 99 | |
AK150291.1 | - | 8 - 9 | 837 | 99 | |
AK152536.1 | - | 8 - 9 | 837 | 99 | |
AK160862.1 | - | 7 - 8 | 686 | 99 | |
AK166534.1 | - | 8 - 9 | 793 | 99 | |
AK167756.1 | - | 8 - 9 | 795 | 99 | |
AK167766.1 | - | 8 - 9 | 826 | 99 | |
AK167862.1 | - | 8 - 9 | 826 | 99 | |
AK167983.1 | - | 8 - 9 | 826 | 99 | |
AK168033.1 | - | 8 - 9 | 795 | 99 | |
AK168162.1 | - | 8 - 9 | 797 | 99 | |
AK168613.1 | - | 8 - 9 | 828 | 99 | |
AK169114.1 | - | 8 - 9 | 790 | 99 | |
AK169335.1 | - | 8 - 9 | 799 | 99 | |
BC049115.1 | - | 8 - 9 | 808 | 99 | |
BC051187.1 | - | 3 - 4 | 385 | 99 | |
BC132547.1 | - | 8 - 9 | 795 | 99 | |
BC138575.1 | - | 8 - 9 | 795 | 99 |