Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017381.2 | NP_059077.1 | 9 - 10 | 952 | 95 |
GenBank mRNA | AK146275.1 | - | 9 - 10 | 950 | 95 |
AK146854.1 | - | 9 - 10 | 944 | 95 | |
AK150291.1 | - | 9 - 10 | 991 | 95 | |
AK152536.1 | - | 9 - 10 | 991 | 95 | |
AK160862.1 | - | 8 - 9 | 840 | 95 | |
AK166534.1 | - | 9 - 10 | 947 | 95 | |
AK167756.1 | - | 9 - 10 | 949 | 95 | |
AK167766.1 | - | 9 - 10 | 980 | 95 | |
AK167862.1 | - | 9 - 10 | 980 | 95 | |
AK167983.1 | - | 9 - 10 | 980 | 95 | |
AK168033.1 | - | 9 - 10 | 949 | 95 | |
AK168162.1 | - | 9 - 10 | 951 | 95 | |
AK168613.1 | - | 9 - 10 | 982 | 95 | |
AK169114.1 | - | 9 - 10 | 944 | 95 | |
AK169335.1 | - | 9 - 10 | 953 | 95 | |
BC049115.1 | - | 9 - 10 | 962 | 95 | |
BC051187.1 | - | 4 - 5 | 539 | 95 | |
BC132547.1 | - | 9 - 10 | 949 | 95 | |
BC138575.1 | - | 9 - 10 | 949 | 95 |