Assay Design
Gene Transcripts
- Gene Symbol
- Ccdc81
- Entrez Gene ID
-
70884
- Gene Name
- coiled-coil domain containing 81
- Gene Aliases
-
4921513D09Rik
- Chromosome Location
-
Chr.7: 89866148 - 89903629 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK029544.1 |
- |
15 - 16 |
2294 |
76 |
AV255663.2 |
- |
4 - 5 |
496 |
76 |
View Full Product Details