Assay Design
Gene Transcripts
- Gene Symbol
- Pgpep1l
- Entrez Gene ID
-
78444
- Gene Name
- pyroglutamyl-peptidase I-like
- Gene Aliases
-
C330024D12Rik
- Chromosome Location
-
Chr.7: 68236450 - 68264484 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_030101.1 |
NP_084377.1 |
1 - 2 |
82 |
80 |
View Full Product Details