Assay Design



Gene Transcripts

Gene Symbol
Alpk3
Entrez Gene ID
116904
Gene Name
alpha-kinase 3
Gene Aliases
AW319487, D330016D04, MAK, Midori, mKIAA1330
Chromosome Location
Chr.7: 81057600 - 81105612 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054085.2 NP_473426.2 6 - 7 3780 104
GenBank mRNA AF338872.1 - 6 - 7 3774 104
AK220384.1 - 2 - 3 2163 104

View Full Product Details