Assay Design
Gene Transcripts
- Gene Symbol
- Ttll13
- Entrez Gene ID
-
269954
- Gene Name
- tubulin tyrosine ligase-like family, member 13
- Gene Aliases
-
1700111A04Rik, 4921538O11, BC024578
- Chromosome Location
-
Chr.7: 80246376 - 80260821 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC145749.1 |
- |
10 - 11 |
1448 |
71 |
View Full Product Details