Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177765.3 | NP_808433.2 | 13 - 14 | 2201 | 77 |
GenBank mRNA | AK029565.1 | - | 13 - 14 | 2200 | 77 |
AM690757.1 | - | 12 - 13 | 1902 | 77 | |
BC145749.1 | - | 12 - 13 | 1809 | 77 |