Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109753.1 | NP_001103223.1 | 9 - 10 | 1927 | 151 |
NM_153579.4 | NP_705807.2 | 9 - 10 | 2134 | 151 | |
GenBank mRNA | AK029736.1 | - | 10 - 11 | 1959 | 151 |
AK043830.1 | - | 9 - 10 | 1854 | 151 | |
AK122359.1 | - | 8 - 9 | 1079 | 151 | |
AK139478.1 | - | 9 - 10 | 1829 | 151 | |
AK140104.1 | - | 9 - 10 | 1847 | 151 | |
AK147261.1 | - | 9 - 10 | 1921 | 151 | |
AK159028.1 | - | 9 - 10 | 1832 | 151 | |
BC060224.1 | - | 8 - 9 | 1518 | 151 | |
BC079568.1 | - | 9 - 10 | 2134 | 151 |