Assay Design
Gene Transcripts
- Gene Symbol
- Qk
- Entrez Gene ID
-
19317
- Gene Name
- quaking
- Gene Aliases
-
1110003F05Rik, 1500005P18, QkI, l(17)-1Wis, l17Wis1
- Chromosome Location
-
Chr.17: 10202856 - 10319361 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_021881.2 |
NP_068681.1 |
6 - 7 |
1422 |
103 |
GenBank mRNA |
U44940.1 |
- |
6 - 7 |
1441 |
103 |
View Full Product Details