Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113358.1 | NP_001106829.1 | 1 - 2 | 291 | 89 |
NM_010015.4 | NP_034145.1 | 1 - 2 | 291 | 89 | |
GenBank mRNA | AK002446.1 | - | 1 - 2 | 277 | 89 |
AK006711.1 | - | 1 - 2 | 222 | 89 | |
AK030925.1 | - | 1 - 2 | 241 | 89 | |
AK054480.1 | - | 1 - 2 | 230 | 89 | |
BC024378.1 | - | 1 - 2 | 224 | 89 | |
BC053379.1 | - | 1 - 2 | 214 | 89 | |
BC058116.1 | - | 1 - 2 | 244 | 89 | |
CN833883.1 | - | - | 417 | 89 | |
U22107.1 | - | 1 - 2 | 204 | 89 | |
U83628.1 | - | 1 - 2 | 234 | 89 |