Assay Design



Gene Transcripts

Gene Symbol
Depdc1a
Entrez Gene ID
76131
Gene Name
DEP domain containing 1a
Gene Aliases
5830484J08Rik, Depdc1
Chromosome Location
Chr.3: 159495426 - 159529955 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172092.1 NP_001165563.1 10 - 11 2183 79
NM_001172093.1 NP_001165564.1 9 - 10 1352 79
NM_029523.3 NP_083799.2 10 - 11 2183 79
GenBank mRNA AK018060.1 - 3 - 4 706 79
AK050456.1 - 9 - 10 1289 79
AK088719.1 - 9 - 10 1328 79
AK146458.1 - 10 - 11 2152 79
BC005799.1 - 3 - 4 506 79
BC023934.1 - 10 - 11 2176 79

View Full Product Details