Assay Design



Gene Transcripts

Gene Symbol
Depdc1a
Entrez Gene ID
76131
Gene Name
DEP domain containing 1a
Gene Aliases
5830484J08Rik, Depdc1
Chromosome Location
Chr.3: 159495426 - 159529955 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172092.1 NP_001165563.1 8 - 9 1824 69
NM_029523.3 NP_083799.2 8 - 9 1824 69
GenBank mRNA AK018060.1 - 1 - 2 347 69
AK146458.1 - 8 - 9 1793 69
BC005799.1 - 1 - 2 147 69
BC023934.1 - 8 - 9 1817 69

View Full Product Details