Assay Design



Gene Transcripts

Gene Symbol
1700007P06Rik
Entrez Gene ID
69308
Gene Name
RIKEN cDNA 1700007P06 gene
Gene Aliases
-
Chromosome Location
Chr.1: 187125138 - 187127852 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_040554.1 1 - 2 180 72
GenBank mRNA AK005744.1 - 1 - 2 181 72


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN836487.1 - 1 - 2 183 Chr. 1 - 187125138 - 187125917 [+] on Build GRCm38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream predicted gene 15509 Gm15509 OTTMUSG00000023012 -103.0 Anti-sense Chr. - 187120856 - 187125035 on Build GRCm38
Downstream G patch domain containing 2 Gpatch2 5830433G22Rik, 5830436K05Rik, AI427714, AI447508, AW107440, AW491060, Gpatc2 89.4kb Sense Chr. - 187215393 - 187369480 on Build GRCm38

View Full Product Details