Assay Design



Gene Transcripts

Gene Symbol
Ak5
Entrez Gene ID
229949
Gene Name
adenylate kinase 5
Gene Aliases
-
Chromosome Location
Chr.3: 152462815 - 152668376 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081277.1 NP_001074746.1 7 - 8 1261 77
GenBank mRNA AK053807.1 - 7 - 8 1261 77

View Full Product Details