Assay Design



Gene Transcripts

Gene Symbol
Zdhhc9
Entrez Gene ID
208884
Gene Name
zinc finger, DHHC domain containing 9
Gene Aliases
6430508G22, 9530098M12Rik
Chromosome Location
Chr.X: 48171967 - 48208702 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172465.4 NP_766053.1 1 - 2 225 59
GenBank mRNA AK032233.1 - 1 - 2 207 59
AK079248.1 - 1 - 2 191 59
BC042618.1 - 1 - 2 84 59

View Full Product Details